Assay Details
Gene Symbol: PADI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009791.1 | NP_001009791.1 | 13 - 14 | 1576 | 59 | |
GenBank mRNA | U85264.1 | - | 13 - 14 | 1576 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
