This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04657815_g1
See other SLC25A19 GE Assays ›
Gene Symbol:
SLC25A19
Gene Name:
solute carrier family 25 member 19
Gene Aliases:
-
Chromosome Location:
Chr.11: 7048212 - 7059721 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04657815_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC25A19
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001142890.1 NP_001136362.1 2 - 3 216 57
XM_012185066.3 XP_012040456.2 3 - 4 320 57
XM_012185067.3 XP_012040457.2 4 - 5 395 57
XM_012185068.3 XP_012040458.2 4 - 5 367 57
XM_012185069.3 XP_012040459.2 3 - 4 292 57
XM_027973932.1 XP_027829733.1 3 - 4 320 57
GenBank mRNA FJ422548.1 - 2 - 3 216 57

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Ovis aries

Panther Classification:

Molecular Function -

mitochondrial carrier protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0