Assay Details
Gene Symbol: CYM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009804.1 | NP_001009804.1 | 4 - 5 | 463 | 77 | |
GenBank mRNA | X53037.1 | - | 463 | 77 |

Gene Symbol: CYM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009804.1 | NP_001009804.1 | 4 - 5 | 463 | 77 | |
GenBank mRNA | X53037.1 | - | 463 | 77 |