Assay Details
| Gene Symbol: CYM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009804.1 | NP_001009804.1 | 7 - 8 | 923 | 68 | |
| GenBank mRNA | X53037.1 | - | 923 | 68 | ||
| Gene Symbol: CYM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009804.1 | NP_001009804.1 | 7 - 8 | 923 | 68 | |
| GenBank mRNA | X53037.1 | - | 923 | 68 | ||