Assay Details
Gene Symbol: GOLGA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017958.4 | XP_004018007.1 | 4 - 5 | 1185 | 68 | |
XM_012098949.3 | XP_011954339.1 | 4 - 5 | 1155 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
