This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa03259225_m1
See other IGF2 GE Assays ›
Gene Symbol:
IGF2
Gene Name:
insulin like growth factor 2
Gene Aliases:
IGF-II
Chromosome Location:
Chr.21: 51230091 - 51254815 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa03259225_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IGF2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001009311.1 NP_001009311.1 3 - 4 439 54
XM_015102891.2 XP_014958377.1 2 - 3 1086 54
XM_027959249.1 XP_027815050.1 3 - 4 394 54
XM_027959250.1 XP_027815051.1 5 - 6 871 54
XM_027959252.1 XP_027815053.1 2 - 3 275 54
XM_027959253.1 XP_027815054.1 4 - 5 668 54
GenBank mRNA HQ711956.1 - 1 - 2 157 54
KU316367.1 - 1 - 2 42 54
M89788.1 - 2 - 3 258 54
M89789.1 - 2 - 3 1094 54
X15248.1 - 169 54
X53554.1 - 385 54
X55638.1 - 439 54

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Ovis aries

Panther Classification:

Molecular Function -

growth factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0