This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa03229353_g1
See other FKBP2 GE Assays ›
Gene Symbol:
FKBP2
Gene Name:
FKBP prolyl isomerase 2
Gene Aliases:
-
Chromosome Location:
Chr.21: 44343908 - 44346857 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa03229353_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FKBP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004019645.3 XP_004019694.2 3 - 4 374 74
XM_004019646.4 XP_004019695.2 3 - 4 1415 74
XM_012102093.2 XP_011957483.2 3 - 4 375 74
XM_012102094.3 XP_011957484.2 3 - 4 306 74
XM_027959527.1 XP_027815328.1 3 - 4 388 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0