This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm99999221_m1
See other CCL4 GE Assays ›
Gene Symbol:
Ccl4
Gene Name:
chemokine (C-C motif) ligand 4
Gene Aliases:
AT744.1, Act-2, MIP-1B, Mip1b, Scya4
Chromosome Location:
Chr.11: 83648624 - 83664683 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm99999221_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ccl4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013652.2 NP_038680.1 1 - 2 155 69
GenBank mRNA AF128218.1 - 1 - 2 128 69
AF128219.1 - 1 - 2 128 69
AK156948.1 - 1 - 2 148 69
AK172472.1 - 1 - 2 148 69
BC119257.1 - 1 - 2 102 8734006 69
BC119259.1 - 1 - 2 102 8734008 69
BY163868.1 - 1 - 2 156 69
M23503.1 - 1 - 2 153 69
M35590.1 - 1 - 2 138 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100001256.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ccl4

Panther Classification:

Molecular Function -

cytokine intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

chemotaxis
inflammatory response
immune response
response to toxic substance
leukocyte chemotaxis
positive regulation of tumor necrosis factor production
cellular response to drug
positive regulation of vascular permeability
negative regulation by host of viral transcription
positive regulation of calcium-mediated signaling
positive regulation of calcium ion transport
positive regulation of natural killer cell chemotaxis
cytokine activity
protein binding
chemokine activity
CCR1 chemokine receptor binding
CCR5 chemokine receptor binding
identical protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100011815.mm.1 NM_013652.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100011815.mm.1 NM_013652.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100011815.mm.1 TC1100001256.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0