This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07307849_s1
See other UGT1A6A GE Assays ›
Gene Symbol:
Ugt1a6a
Gene Name:
UDP glucuronosyltransferase 1 family, polypeptide A6A
Gene Aliases:
UDPGT, UDPGT 1-6, UGT1-06, UGT1.6, Ugt1a6, Ugt1a7
Chromosome Location:
Chr.1: 88134809 - 88220002 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07307849_s1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ugt1a6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145079.3 NP_659545.2 418 91
GenBank mRNA AK162514.1 - 414 91
BC012716.1 - 412 91
BC069940.1 - 408 5249697 91
D87867.1 - 1 290 91
U09930.1 - 1 300 91
U16818.1 - 418 91

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ugt1a6a

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
flavonoid biosynthetic process
glucuronate metabolic process
cellular response to hormone stimulus
flavonoid glucuronidation
xenobiotic glucuronidation
glucuronosyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
transferase activity, transferring hexosyl groups
enzyme binding
protein complex binding
protein homodimerization activity
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0