This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07305590_m1
See other DUPD1 GE Assays ›
Gene Symbol:
Dupd1
Gene Name:
dual specificity phosphatase and pro isomerase domain containing 1
Gene Aliases:
EG435391
Chromosome Location:
Chr.14: 21676209 - 21714576 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07305590_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dupd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001013826.2 NP_001013848.1 1 - 2 66 84
GenBank mRNA AK075665.1 - 1 - 2 67 84
BC147517.1 - 1 - 2 29 9056509 84
BC147531.1 - 1 - 2 29 9056522 84

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK075665.1, BC147517.1, BC147531.1, NM_001013826.2 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
dephosphorylation
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
protein tyrosine/serine/threonine phosphatase activity
hydrolase activity
phosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0