This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07301737_m1
See other IDH2 GE Assays ›
Gene Symbol:
Idh2
Gene Name:
isocitrate dehydrogenase 2 (NADP+), mitochondrial
Gene Aliases:
E430004F23, IDPm, Idh-2
Chromosome Location:
Chr.7: 80094846 - 80115387 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07301737_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Idh2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_173011.2 NP_766599.2 8 - 9 1153 68
GenBank mRNA AF212319.1 - 8 - 9 1124 68
AK088110.1 - 8 - 9 1125 68
AK145753.1 - 8 - 9 1098 68
AK146564.1 - 8 - 9 1239 68
AK161640.1 - 8 - 9 1137 68
AK169395.1 - 8 - 9 1150 68
BC060030.1 - 8 - 9 1141 6515657 68

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Idh2

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

glyoxylate cycle
tricarboxylic acid cycle
isocitrate metabolic process
oxidation-reduction process
magnesium ion binding
isocitrate dehydrogenase (NADP+) activity
oxidoreductase activity
oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
metal ion binding
NAD binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0