This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07300404_m1
See other PDE6A GE Assays ›
Gene Symbol:
Pde6a
Gene Name:
phosphodiesterase 6A, cGMP-specific, rod, alpha
Gene Aliases:
Pdea, nmf282
Chromosome Location:
Chr.18: 61220080 - 61289750 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07300404_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pde6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_146086.2 NP_666198.1 9 - 10 1366 87
GenBank mRNA BC031925.1 - 9 - 10 1366 4501123 87
BC044892.1 - 9 - 10 1366 4527430 87
BC158126.1 - 9 - 10 1550 9007394 87
X60664.1 - 9 - 10 1374 87

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pde6a

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
visual perception
photoreceptor cell maintenance
GMP metabolic process
response to stimulus
regulation of cytosolic calcium ion concentration
retina development in camera-type eye
3',5'-cyclic-nucleotide phosphodiesterase activity
phosphoric diester hydrolase activity
hydrolase activity
metal ion binding
3',5'-cyclic-GMP phosphodiesterase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0