This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07300328_s1
See other RGS9BP GE Assays ›
Gene Symbol:
Rgs9bp
Gene Name:
regulator of G-protein signalling 9 binding protein
Gene Aliases:
R9AP, Rgs9-1bp
Chromosome Location:
Chr.7: 35578993 - 35585582 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07300328_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rgs9bp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145840.3 NP_665839.1 1 823 77
GenBank mRNA AF483907.1 - 1 461 77
AK044285.1 - 1 824 77
AY229888.1 - 1 785 77
BC118006.1 - 1 779 40081388 77

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rgs9bp

Gene Ontology Categories:

Function(s)

visual perception
negative regulation of signal transduction
response to stimulus
detection of light stimulus involved in visual perception

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-867854cbc7-vwcfx:80/100.66.130.94:80.
git-commit: 25ccc7b1e9fd8ebf0690872e1fef11e169411d8b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0