This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07299570_m1
See other N4BP1 GE Assays ›
Gene Symbol:
N4bp1
Gene Name:
NEDD4 binding protein 1
Gene Aliases:
AI481586, C81621
Chromosome Location:
Chr.8: 86841136 - 86885258 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07299570_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: N4bp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_030563.2 NP_085040.2 3 - 4 2252 93
GenBank mRNA AK172981.1 - 3 - 4 2234 93
BC089324.1 - 2 - 3 1417 93

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene N4bp1

Panther Classification:

Molecular Function -

endoribonuclease

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of protein ubiquitination
negative regulation of proteasomal ubiquitin-dependent protein catabolic process
cellular response to UV
RNA binding
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0