This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07298985_g1
See other MAGEB16 GE Assays ›
Gene Symbol:
Mageb16
Gene Name:
melanoma antigen family B, 16
Gene Aliases:
2410003J06Rik
Chromosome Location:
Chr.X: 79623231 - 79671435 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07298985_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mageb16
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001113734.1 NP_001107206.1 2 - 3 170 72
NM_028025.1 NP_082301.1 3 - 4 299 72
GenBank mRNA AK010362.1 - 3 - 4 300 72
AK167347.1 - 2 - 3 143 72
BC050207.1 - 2 - 3 154 30007513 72
BC115695.1 - 2 - 3 130 40104521 72
CJ081836.1 - 2 - 3 171 72

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK010362.1, AK167347.1, BC050207.1, BC115695.1, NM_001113734.1, NM_028025.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0