This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07298495_m1
See other SLC47A1 GE Assays ›
Gene Symbol:
Slc47a1
Gene Name:
solute carrier family 47, member 1
Gene Aliases:
1300013J15Rik, AI036982, MATE1, mMATE1
Chromosome Location:
Chr.11: 61343400 - 61378781 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07298495_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc47a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026183.5 NP_080459.2 3 - 4 328 72
GenBank mRNA AK004994.2 - 3 - 4 318 72
BB661673.1 - 3 - 4 325 72
BC031436.1 - 3 - 4 312 72

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK004994.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc47a1

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
drug transmembrane transport
organic cation transport
drug transport
transmembrane transport
hydrogen ion transmembrane transport
monovalent cation:proton antiporter activity
drug transmembrane transporter activity
antiporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0