This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07297345_m1
See other HUNK GE Assays ›
Gene Symbol:
Hunk
Gene Name:
hormonally upregulated Neu-associated kinase
Gene Aliases:
Bstk1, Mak-v
Chromosome Location:
Chr.16: 90385797 - 90499555 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07297345_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hunk
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_015755.2 NP_056570.1 4 - 5 810 70
GenBank mRNA AF055919.2 - 4 - 5 810 70
AF167987.1 - 4 - 5 822 70
AK147425.1 - 4 - 5 1195 70
AK147734.1 - 4 - 5 564 70
BC137788.1 - 4 - 5 871 8860807 70
BC137790.1 - 4 - 5 870 8860809 70

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hunk

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
phosphorylation
intracellular signal transduction
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0