This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07296658_m1
See other GBP2B GE Assays ›
Gene Symbol:
Gbp2b
Gene Name:
guanylate binding protein 2b
Gene Aliases:
Gbp-1, Gbp1, Mag-1, Mpa-1, Mpa1
Chromosome Location:
Chr.3: 142594816 - 142619179 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07296658_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gbp2b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010259.2 NP_034389.2 5 - 6 702 98
GenBank mRNA AK077796.1 - 5 - 6 703 98
BC108990.2 - 747 40047769 98
EF494422.1 - 5 - 6 702 98
M55544.1 - 686 98
M63961.1 - 780 98

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gbp2b

Panther Classification:

Molecular Function -

heterotrimeric G-protein

Gene Ontology Categories:

Function(s) Process(es)

immune system process
cellular response to interferon-beta
defense response to protozoan
adhesion of symbiont to host
defense response to Gram-positive bacterium
defense response to virus
cellular response to interferon-gamma
nucleotide binding
GTPase activity
GTP binding
GMP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0