This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07296508_g1
See other CDK4 GE Assays ›
Gene Symbol:
Cdk4
Gene Name:
cyclin-dependent kinase 4
Gene Aliases:
Crk3
Chromosome Location:
Chr.10: 127063537 - 127067288 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07296508_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdk4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009870.3 NP_034000.1 1 - 2 157 59
GenBank mRNA AK003804.1 - 1 - 2 124 59
AK006918.1 - 1 - 2 124 59
AK011871.1 - 1 - 2 145 59
AK012064.1 - 1 - 2 127 59
AK019380.1 - 1 - 2 111 59
AK078440.1 - 1 - 2 111 59
AK146107.1 - 1 - 2 124 59
AK153465.1 - 1 - 2 157 59
AK185090.1 - 1 - 2 47 59
BC031599.1 - 1 - 2 146 59
BC046336.1 - 1 - 2 157 6312184 59
BC052694.1 - 1 - 2 152 5714971 59

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK003804.1, AK011871.1, AK078440.1, AK146107.1, AK153465.1, BC046336.1, BC052694.1, NM_009870.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cdk4

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
protein phosphorylation
cell cycle
signal transduction
positive regulation of cell proliferation
response to organic substance
response to lead ion
regulation of gene expression
positive regulation of G2/M transition of mitotic cell cycle
phosphorylation
response to testosterone
regulation of cell proliferation
response to drug
positive regulation of apoptotic process
positive regulation of translation
positive regulation of cell size
positive regulation of fibroblast proliferation
cell division
regulation of cell cycle
negative regulation of cell cycle arrest
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
cyclin-dependent protein serine/threonine kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
cyclin binding
protein complex binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0