This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07295928_m1
See other PTGDR GE Assays ›
Gene Symbol:
Ptgdr
Gene Name:
prostaglandin D receptor
Gene Aliases:
DP, PGD
Chromosome Location:
Chr.14: 44851235 - 44859444 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07295928_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ptgdr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008962.4 NP_032988.3 1 - 2 963 105
GenBank mRNA AK032488.1 - 1 - 2 964 105
BC132297.1 - 1 - 2 942 40130574 105

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ptgdr

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

inflammatory response
signal transduction
G-protein coupled receptor signaling pathway
male sex determination
sleep
adenosine metabolic process
cellular response to prostaglandin D stimulus
prostaglandin J receptor activity
signal transducer activity
G-protein coupled receptor activity
prostaglandin D receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0