This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07295531_g1
See other CALM2 GE Assays ›
Gene Symbol:
Calm2
Gene Name:
calmodulin 2
Gene Aliases:
1500001E21Rik, AL024017
Chromosome Location:
Chr.17: 87433401 - 87446935 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07295531_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Calm2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007589.5 NP_031615.1 4 - 5 428 91
GenBank mRNA AK002974.1 - 389 91
AK012247.1 - 4 - 5 356 91
AK012564.1 - 4 - 5 357 91
AK013068.1 - 4 - 5 350 91
AK152303.1 - 4 - 5 360 91
AK160636.1 - 4 - 5 363 91
AK161268.1 - 4 - 5 356 91
AK161984.1 - 4 - 5 357 91
AK168741.1 - 4 - 5 357 91
BC010730.1 - 2 - 3 133 91
BC021347.1 - 4 - 5 359 4006686 91
BC051444.1 - 4 - 5 352 30072702 91
BC100301.1 - 4 - 5 343 30526959 91

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Calm2

Panther Classification:

Molecular Function -

calmodulin-related

Gene Ontology Categories:

Function(s) Process(es)

G2/M transition of mitotic cell cycle
regulation of catalytic activity
calcium ion binding
adenylate cyclase binding
protein domain specific binding
enzyme regulator activity
nitric-oxide synthase regulator activity
type 3 metabotropic glutamate receptor binding
phosphatidylinositol 3-kinase binding
ion channel binding
protein N-terminus binding
calcium-dependent protein binding
nitric-oxide synthase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0