This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07295344_m1
See other MPZL1 GE Assays ›
Gene Symbol:
Mpzl1
Gene Name:
myelin protein zero-like 1
Gene Aliases:
1110007A10Rik, PZR
Chromosome Location:
Chr.1: 165592181 - 165634558 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07295344_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mpzl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001001880.2 NP_001001880.1 1 - 2 236 104
NM_001083897.1 NP_001077366.1 1 - 2 236 104
GenBank mRNA AK090278.1 - 1 - 2 223 104
AK169385.1 - 1 - 2 226 104
AY764247.1 - 1 - 2 233 104
AY764248.1 - 1 - 2 88 104
BB871672.1 - 1 - 2 236 104
BC072563.1 - 1 - 2 231 6808882 104

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mpzl1

Panther Classification:

Molecular Function -

myelin protein

Gene Ontology Categories:

Function(s)

signal transduction

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0