This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07295300_g1
See other LUC7L2 GE Assays ›
Gene Symbol:
Luc7l2
Gene Name:
LUC7-like 2 (S. cerevisiae)
Gene Aliases:
4930471C18Rik, AA522013, AU015269, CGI59, Cgi74
Chromosome Location:
Chr.6: 38551339 - 38609471 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07295300_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Luc7l2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK140271.1 - 3 - 4 2519 88
AK147889.1 - 3 - 4 561 88
AK153527.1 - 3 - 4 600 88

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK140271.1, AK147889.1, AK153527.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0