This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07294219_m1
See other GSTA3 GE Assays ›
Gene Symbol:
Gsta3
Gene Name:
glutathione S-transferase, alpha 3
Gene Aliases:
Gst2-3
Chromosome Location:
Chr.1: 21240585 - 21265575 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07294219_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gsta3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001077353.2 NP_001070821.1 4 - 5 334 65
NM_001288617.1 NP_001275546.1 4 - 5 379 65
NM_010356.4 NP_034486.2 3 - 4 239 65
GenBank mRNA AI047326.1 - 4 - 5 312 65
AI255699.1 - 3 - 4 235 65
AK002478.1 - 2 - 3 187 65
AK014076.1 - 3 - 4 212 65
AK149543.1 - 3 - 4 204 65
BC147272.1 - 3 - 4 202 9056327 65
BC147273.1 - 3 - 4 202 9056339 65
GQ227791.1 - 2 - 3 164 65
M73483.1 - 2 - 3 170 65
X65021.1 - 2 - 3 155 65

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001288617.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gsta3

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
aflatoxin catabolic process
glutathione transferase activity
transferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0