This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07294087_m1
See other UBE2T GE Assays ›
Gene Symbol:
Ube2t
Gene Name:
ubiquitin-conjugating enzyme E2T (putative)
Gene Aliases:
2700084L22Rik, C80607
Chromosome Location:
Chr.1: 134962565 - 134974180 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07294087_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ube2t
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001278115.1 NP_001265044.1 3 - 4 281 67
NM_026024.3 NP_080300.1 3 - 4 458 67
GenBank mRNA AK012565.1 - 3 - 4 417 67
AK021213.1 - 3 - 4 447 67
AK166098.1 - 3 - 4 428 67
BC029213.1 - 3 - 4 407 3591557 67
CK334733.1 - 3 - 4 272 67

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ube2t

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
DNA repair
protein monoubiquitination
cellular response to DNA damage stimulus
protein K29-linked ubiquitination
protein K27-linked ubiquitination
protein autoubiquitination
protein K63-linked ubiquitination
protein K48-linked ubiquitination
protein K11-linked ubiquitination
protein K6-linked ubiquitination
nucleotide binding
chromatin binding
ubiquitin-protein transferase activity
ATP binding
ligase activity
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0