This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07294065_m1
See other CRKL GE Assays ›
Gene Symbol:
Crkl
Gene Name:
v-crk sarcoma virus CT10 oncogene homolog (avian)-like
Gene Aliases:
1110025F07Rik, AA589403, AI325100, Crkol
Chromosome Location:
Chr.16: 17451985 - 17487440 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07294065_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Crkl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001277231.1 NP_001264160.1 1 - 2 809 84
GenBank mRNA CA328740.1 - 1 - 2 318 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

blood vessel development
pattern specification process
heart development
organ morphogenesis
anterior/posterior pattern specification
thymus development
parathyroid gland development
protein binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0