This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07293712_g1
See other DDX39B GE Assays ›
Gene Symbol:
Ddx39b
Gene Name:
DEAD (Asp-Glu-Ala-Asp) box polypeptide 39B
Gene Aliases:
0610030D10Rik, AI428441, Bat-1, Bat1, Bat1a, D17H6S81E, D17H6S81E-1, D6S81Eh
Chromosome Location:
Chr.17: 35241746 - 35253707 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07293712_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ddx39b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252457.1 NP_001239386.1 5 - 6 760 100
NM_019693.3 NP_062667.1 5 - 6 848 100
GenBank mRNA AF118128.1 - 4 - 5 614 100
AK051034.1 - 5 - 6 781 100
AK088867.1 - 5 - 6 766 100
AK171507.1 - 5 - 6 790 100
AK194985.1 - 1 - 2 165 100
AK217230.1 - 1 - 2 135 100
AY255786.1 - 4 - 5 740 100
AY255787.1 - 4 - 5 740 100
BC011067.1 - 5 - 6 756 3498520 100
BC024859.1 - 4 - 5 1984 5359825 100

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ddx39b

Gene Ontology Categories:

Function(s) Process(es)

spliceosomal complex assembly
mRNA splicing, via spliceosome
mRNA processing
mRNA export from nucleus
transport
cellular response to DNA damage stimulus
RNA splicing
regulation of gene expression
RNA secondary structure unwinding
positive regulation of DNA-templated transcription, elongation
viral mRNA export from host cell nucleus
mRNA transport
negative regulation of DNA damage checkpoint
nucleotide binding
nucleic acid binding
RNA binding
ATP-dependent RNA helicase activity
helicase activity
ATP binding
RNA-dependent ATPase activity
hydrolase activity
U6 snRNA binding
U4 snRNA binding
ATP-dependent protein binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0