This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm06660894_s1
See other P4HB GE Assays ›
Gene Symbol:
P4hb
Gene Name:
prolyl 4-hydroxylase, beta polypeptide
Gene Aliases:
ERp59, PDI, Pdia1, Thbp
Chromosome Location:
Chr.11: 120560304 - 120572936 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm06660894_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: P4hb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011032.2 NP_035162.1 11 1759 87
GenBank mRNA AK141304.1 - 11 88
AK146542.1 - 11 1697 87
AK150002.1 - 11 1723 87
AK150422.1 - 11 1697 87
AK150592.1 - 11 1696 87
AK150755.1 - 11 1683 87
AK150805.1 - 11 1697 87
AK151351.1 - 11 1691 87
AK151546.1 - 11 1697 87
AK152141.1 - 11 1697 87
AK152217.1 - 11 1697 87
AK152791.1 - 11 1696 87
AK152834.1 - 11 1697 87
AK153365.1 - 11 1696 87
AK159606.1 - 11 1697 87
AK159729.1 - 11 1697 87
AK159965.1 - 11 1697 87
AK167757.1 - 4 535 87
AK167796.1 - 11 1697 87
AK168330.1 - 11 1695 87
AK168336.1 - 11 1679 87
AK168612.1 - 11 1697 87
AK168893.1 - 11 1697 87
AK169265.1 - 11 1694 87
AK170603.1 - 11 1694 87
BC008549.1 - 11 1685 3591404 87
BC018581.1 - 4 545 87
BC093512.1 - 11 1676 5124376 87
J05185.1 - 11 1759 87
X06453.1 - 11 1699 87

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene P4hb

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein folding
peptidyl-proline hydroxylation to 4-hydroxy-L-proline
response to endoplasmic reticulum stress
cell redox homeostasis
cellular response to hypoxia
regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
protein disulfide isomerase activity
procollagen-proline 4-dioxygenase activity
isomerase activity
enzyme binding
peptidyl-proline 4-dioxygenase activity
poly(A) RNA binding
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0