This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm06658056_g1
See other RPL5 GE Assays ›
Gene Symbol:
Rpl5 Fam69a
Gene Name:
ribosomal protein L5
family with sequence similarity 69, member A
Gene Aliases:
U21RNA
2900024C23Rik, AI315274
Chromosome Location:
Chr.5: 107900528 - 107909005 on Build GRCm38
Chromosome Location:
Chr.5: 107908042 - 107987077 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
Assay ID Mm06658056_g1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rpl5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_016980.2 NP_058676.1 8 972 95
GenBank mRNA AK008489.1 - 8 933 95
AK013107.1 - 8 930 95
AK032778.1 - 8 930 95
AK088036.1 - 8 932 95
AK152381.1 - 8 929 95
AK166865.1 - 9 888 95
AK168317.1 - 4 377 95
AK169313.1 - 8 930 95
BC005790.1 - 8 911 95
BC026934.1 - 8 910 2651564 95
BC083318.1 - 8 912 6818756 95
BC091752.1 - 8 910 4208431 95
BC106128.1 - 9 877 2651407 95
CV561106.1 - 2 115 95
X83590.1 - 3 241 95
Gene Symbol: Fam69a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA DT926079.1 - 1 275 95

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rpl5
Click here to find siRNAs for Gene Fam69a

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

rRNA processing
translation
positive regulation of phosphatase activity
ribosomal large subunit biogenesis
RNA binding
structural constituent of ribosome
protein binding
5S rRNA binding
rRNA binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0