This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm06654625_g1
See other CABP4 GE Assays ›
Gene Symbol:
Cabp4
Gene Name:
calcium binding protein 4
Gene Aliases:
2410038D05Rik
Chromosome Location:
Chr.19: 4135423 - 4144080 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm06654625_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cabp4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144532.2 NP_653115.1 5 826 74
GenBank mRNA AK010631.1 - 2 368 74
AK171863.1 - 8 2768 74
AY039218.1 - 5 821 74
BC029002.1 - 4 2066 74
BC049263.1 - 5 826 5366999 74
BC053330.1 - 4 2113 74

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Amplicon spans exons and probe does not span exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cabp4

Gene Ontology Categories:

Function(s) Process(es)

visual perception
phototransduction
photoreceptor cell morphogenesis
retinal cone cell development
retinal bipolar neuron differentiation
calcium ion binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0