This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05901949_s1
See other RFK GE Assays ›
Gene Symbol:
Rfk
Gene Name:
riboflavin kinase
Gene Aliases:
0610038L10Rik, AF031381, KOI-4
Chromosome Location:
Chr.19: 17394043 - 17401349 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05901949_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rfk
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019437.3 NP_062310.1 4 1773 107
GenBank mRNA AK002806.1 - 4 1734 107
BC033521.1 - 4 1675 4527980 107
BC051021.1 - 4 1684 6315425 107

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rfk

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
riboflavin biosynthetic process
phosphorylation
positive regulation of NAD(P)H oxidase activity
reactive oxygen species metabolic process
nucleotide binding
protein binding
ATP binding
riboflavin kinase activity
kinase activity
transferase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0