This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05882202_s1
See other PPP1R15B GE Assays ›
Gene Symbol:
Ppp1r15b
Gene Name:
protein phosphatase 1, regulatory (inhibitor) subunit 15b
Gene Aliases:
1810033K10Rik, AI606441, C530022L24Rik, CReP
Chromosome Location:
Chr.1: 133131166 - 133139800 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05882202_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp1r15b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133819.3 NP_598580.1 1 100 97
GenBank mRNA AK084736.1 - 97

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_133819.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp1r15b

Gene Ontology Categories:

Function(s) Process(es)

regulation of translation
regulation of translational initiation
response to oxidative stress
ER overload response
response to endoplasmic reticulum stress
response to hydrogen peroxide
peptidyl-serine dephosphorylation
negative regulation of endoplasmic reticulum stress-induced eIF2 alpha phosphorylation
protein serine/threonine phosphatase activity
translation regulator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0