This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05879026_gH
See other CCRN4L GE Assays ›
Gene Symbol:
Ccrn4l BC094435 Sgip1 Slc38a6
Gene Name:
CCR4 carbon catabolite repression 4-like (S. cerevisiae)
cDNA sequence BC094435
SH3-domain GRB2-like (endophilin) interacting protein 1
solute carrier family 38, member 6
Gene Aliases:
AU043840, Ccr4, nocturnin
ENSMUSG00000073624
3110007P09Rik, SGIP1alpha, gag
AW322671, EG625098
Chromosome Location:
Chr.3: 51224447 - 51251654 on Build GRCm38
Chromosome Location:
-
Chromosome Location:
Chr.4: 102759167 - 102972701 on Build GRCm38
Chromosome Location:
Chr.12: 73286794 - 73361998 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
-
-
Assay ID Mm05879026_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ccrn4l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC026940.1 - 1543 117
Gene Symbol: BC094435
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK088963.1 - 1 1139 117
BC094435.1 - 1 1173 117
Gene Symbol: Sgip1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK090117.1 - 4167 117
AK138403.1 - 1224 117
Gene Symbol: Slc38a6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK054341.1 - 5 117

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

viral or transposable element protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0