This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05873489_s1
See other DPP3 GE Assays ›
Gene Symbol:
Dpp3
Gene Name:
dipeptidylpeptidase 3
Gene Aliases:
4930533O14Rik, C86324
Chromosome Location:
Chr.19: 4907229 - 4928287 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05873489_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dpp3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133803.2 NP_598564.2 18 2382 113
GenBank mRNA AK031020.1 - 18 2347 113
AK087994.1 - 18 113
AK150101.1 - 18 2342 113
AK150126.1 - 18 2336 113
AK166842.1 - 18 2347 113
AK168195.1 - 18 113
AK218694.1 - 1 203 113
BC004600.1 - 17 2324 3584924 113
BC066817.1 - 18 2333 30429618 113

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dpp3

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
aminopeptidase activity
peptidase activity
metallopeptidase activity
dipeptidyl-peptidase activity
zinc ion binding
hydrolase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0