Assay Details
Gene Symbol: Igfbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK045003.1 | - | 1 | 1226 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Gene Symbol: Igfbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK045003.1 | - | 1 | 1226 | 97 |
Set Membership: |
![]() ![]() ![]() ![]() |