This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05804785_gH
See other AKT2 GE Assays ›
Gene Symbol:
Akt2
Gene Name:
thymoma viral proto-oncogene 2
Gene Aliases:
2410016A19Rik, AW554154, PKB, PKBbeta
Chromosome Location:
Chr.7: 27591557 - 27639453 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05804785_gH
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Akt2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001110208.1 NP_001103678.1 2 - 3 447 59
NM_007434.3 NP_031460.1 2 - 3 205 59
GenBank mRNA AK144217.1 - 2 - 3 264 59
AK158792.1 - 2 - 3 448 59
BC026151.1 - 2 - 3 228 4187425 59
BC028759.1 - 1 - 2 121 59
BC040377.1 - 2 - 3 197 4500854 59
U22445.1 - 2 - 3 258 59
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK050325.1 - 758 Chr. - - [-] on Build 59

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Akt2

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein phosphorylation
carbohydrate metabolic process
glycogen metabolic process
glycogen biosynthetic process
glucose metabolic process
regulation of translation
protein phosphorylation
transport
apoptotic process
multicellular organism development
insulin receptor signaling pathway
carbohydrate transport
positive regulation of signal transduction
positive regulation of gene expression
negative regulation of plasma membrane long-chain fatty acid transport
positive regulation of sodium ion transport
positive regulation of glucose metabolic process
phosphorylation
positive regulation of cell migration
positive regulation of vesicle fusion
positive regulation of fatty acid beta-oxidation
peripheral nervous system myelin maintenance
cellular response to insulin stimulus
negative regulation of RNA splicing
positive regulation of peptidyl-serine phosphorylation
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
protein kinase B signaling
positive regulation of nitric oxide biosynthetic process
positive regulation of glycogen biosynthetic process
positive regulation of transcription from RNA polymerase II promoter
positive regulation of glucose import
positive regulation of positive chemotaxis
intracellular protein transmembrane transport
cellular response to organic cyclic compound
protein localization to plasma membrane
positive regulation of protein targeting to membrane
activation of GTPase activity
positive regulation of cell motility
positive regulation of glucose import in response to insulin stimulus
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
protein kinase C binding
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0