This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05715849_s1
See other SEPT11 GE Assays ›
Gene Symbol:
Sept11
Gene Name:
septin 11
Gene Aliases:
6230410I01Rik, AW548875, D5Ertd606e
Chromosome Location:
Chr.5: 93091879 - 93179054 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05715849_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sept11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001009818.1 NP_001009818.1 10 2558 67
GenBank mRNA AK136162.1 - 4 1543 67
AK147983.1 - 1 67
AK166517.1 - 10 2580 67
AK170945.1 - 9 2368 67
BC031456.1 - 10 2465 4037515 67
BC064466.1 - 10 2558 6825549 67

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sept11

Panther Classification:

Molecular Function -

cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
protein heterooligomerization
cell division
nucleotide binding
molecular_function
GTP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0