This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05709679_s1
See other RAD1 GE Assays ›
Gene Symbol:
Rad1
Gene Name:
RAD1 homolog (S. pombe)
Gene Aliases:
mRAD1
Chromosome Location:
Chr.15: 10486018 - 10499063 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05709679_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rad1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289447.1 NP_001276376.1 6 891 98
NM_011232.3 NP_035362.2 6 1103 98
GenBank mRNA AF038841.1 - 6 1014 98
AF058394.1 - 5 790 98
AF073523.1 - 6 1030 98
AF074718.1 - 5 800 98
AF076842.1 - 5 801 98
AF084514.1 - 6 962 98
AJ004976.1 - 5 812 98
AK146533.1 - 6 1073 98
AK148098.1 - 6 1062 98
AK150466.1 - 6 1012 98
AK164168.1 - 3 3801 98
BC048693.1 - 5 867 6508142 98

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rad1

Panther Classification:

Molecular Function -

DNA metabolism protein exodeoxyribonuclease nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

DNA damage checkpoint
DNA repair
cellular response to DNA damage stimulus
meiotic recombination checkpoint
cellular response to ionizing radiation
nucleic acid phosphodiester bond hydrolysis
damaged DNA binding
nuclease activity
exonuclease activity
3'-5' exonuclease activity
exodeoxyribonuclease III activity
hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0