This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm05678822_g1
See other PGAM1 GE Assays ›
Gene Symbol:
Pgam1
Gene Name:
phosphoglycerate mutase 1
Gene Aliases:
2310050F24Rik, Pgam-1
Chromosome Location:
Chr.19: 41911871 - 41918665 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm05678822_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pgam1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_023418.2 NP_075907.2 4 897 66
GenBank mRNA AF283667.1 - 4 809 66
AK004921.1 - 4 753 66
AK009905.2 - 4 750 66
AK144681.1 - 4 751 66
AK150716.1 - 4 746 66
AK152443.1 - 4 746 66
AK153202.1 - 4 750 66
AK159305.1 - 4 750 66
AK165410.1 - 4 746 66
AK168949.1 - 4 743 66
AK215282.1 - 2 169 66
AK218196.1 - 2 169 66
AK219760.1 - 2 169 66
BC002241.1 - 4 845 3491964 66
BC005661.1 - 4 811 3588761 66
BC066844.1 - 4 734 30471755 66
BC067412.1 - 3 550 66
BC083090.1 - 4 746 30444229 66
BC106139.1 - 4 817 6528521 66

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pgam1

Panther Classification:

Molecular Function -

mutase

Gene Ontology Categories:

Function(s) Process(es)

glycolytic process
regulation of glycolytic process
metabolic process
regulation of pentose-phosphate shunt
respiratory burst
catalytic activity
bisphosphoglycerate mutase activity
bisphosphoglycerate 2-phosphatase activity
phosphoglycerate mutase activity
hydrolase activity
isomerase activity
intramolecular transferase activity, phosphotransferases
protein kinase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0