This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04934843_m1
See other ZFP950 GE Assays ›
Gene Symbol:
Zfp950
Gene Name:
zinc finger prtoein 950
Gene Aliases:
5830428H23Rik, AU016073, BC055757, Gm34518, Zfp826, mszf84
Chromosome Location:
Chr.19: 61051147 - 61313507 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04934843_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zfp950
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK039536.1 - 3 - 4 324 115
BC086681.1 - 3 - 4 284 115
CV562489.1 - 3 - 4 302 115
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CJ112202.1 - 3 - 4 276 Chr. 19 - 61117164 - 61140793 [-] on Build GRCm38 115
GenBankmRNA CJ110503.1 - 3 - 4 278 Chr. 19 - 61117193 - 61140795 [-] on Build GRCm38 115
GenBankmRNA CJ106654.1 - 3 - 4 278 Chr. 19 - 61117180 - 61140795 [-] on Build GRCm38 115
GenBankmRNA CJ124692.1 - 3 - 4 278 Chr. 19 - 61117182 - 61140795 [-] on Build GRCm38 115
GenBankmRNA CJ060331.1 - 3 - 4 298 Chr. 19 - 61118222 - 61140814 [-] on Build GRCm38 115
GenBankmRNA BB633523.1 - 3 - 4 324 Chr. 19 - 61117054 - 61140840 [-] on Build GRCm38 115
GenBankmRNA CO424014.1 - 3 - 4 284 Chr. 19 - 61116905 - 61140801 [-] on Build GRCm38 115

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream G protein-coupled receptor kinase 5 Grk5 Gprk5 -22.3kb Anti-sense Chr. 19 - 60889593 - 61095968 on NCBI Build GRCm38
Downstream predicted gene 6020 Gm6020 EG547091,
mszf72
24.8kb Anti-sense Chr. 19 - 61151864 - 61184658 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0