This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04934839_g1
See other KIF16BOS GE Assays ›
Gene Symbol:
Kif16bos
Gene Name:
kinesin family member 16B, opposite strand
Gene Aliases:
4930511F01Rik
Chromosome Location:
Chr.2: 142902290 - 142904076 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04934839_g1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Kif16bos
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK015749.1 - 1 - 2 276 164

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream kinesin family member 16B Kif16b 8430434E15Rik,
AA623607,
AI789011,
C80253,
C80902,
mKIAA1590
-1.4kb Anti-sense Chr. 2 - 142618334 - 142901595 on NCBI Build GRCm38
Downstream U2 small nuclear ribonucleoprotein B Snrpb2 2810052G09Rik 159.5kb Sense Chr. 2 - 143063069 - 143072052 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0