This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04933317_g1
See other HAX1 GE Assays ›
Gene Symbol:
Hax1
Gene Name:
HCLS1 associated X-1
Gene Aliases:
HAX-1, HSP1BP-1, Hs1bp1, SIG-111, Silg111, mHAX-1s
Chromosome Location:
Chr.3: 89995446 - 89998716 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04933317_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hax1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011826.4 NP_035956.1 3 - 4 646 97
GenBank mRNA AK002256.1 - 3 - 4 597 97
AK010633.1 - 3 - 4 630 97
AK088746.1 - 3 - 4 628 97
BC006688.1 - 3 - 4 575 3582652 97
BC098225.1 - 3 - 4 600 1494110 97
BC147481.1 - 3 - 4 577 9056226 97

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hax1

Gene Ontology Categories:

Function(s) Process(es)

cell surface receptor signaling pathway
positive regulation of phosphatidylinositol 3-kinase signaling
regulation of actin filament polymerization
positive regulation of granulocyte differentiation
positive regulation of peptidyl-serine phosphorylation
negative regulation of apoptotic process
positive regulation of transcription from RNA polymerase II promoter
positive regulation of peptidyl-tyrosine phosphorylation
positive regulation of protein kinase B signaling
cellular response to cytokine stimulus
positive regulation of actin cytoskeleton reorganization
protein binding
protein domain specific binding
interleukin-1 binding
protein N-terminus binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0