This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04411709_m1
See other GM32014 GE Assays ›
Gene Symbol:
Gm32014
Gene Name:
predicted gene, 32014
Gene Aliases:
-
Chromosome Location:
Chr.9: 8644078 - 8740988 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04411709_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gm32014
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110513.1 4 - 5 582 95
GenBank mRNA AK054106.1 - 4 - 5 582 95

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 1899 Mir1899 Mirn1899,
mmu-mir-1899
-397.2kb Anti-sense Chr. 9 - 8246786 - 8246881 on NCBI Build GRCm38
Downstream progesterone receptor Pgr 9930019P03Rik,
BB114106,
ENSMUSG00000074510,
NR3C3,
PR,
PR-A,
PR-B
205.2kb Anti-sense Chr. 9 - 8894736 - 8968611 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0