This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04411033_m1
See other TERF2 GE Assays ›
Gene Symbol:
Terf2
Gene Name:
telomeric repeat binding factor 2
Gene Aliases:
TRF2
Chromosome Location:
Chr.8: 107069400 - 107096642 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04411033_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Terf2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_104410.1 5 - 6 881 87
GenBank mRNA AK160421.1 - 5 - 6 526 87
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY707010.1 - 5 - 6 524 Chr. 8 - 107079409 - 107096188 [-] on Build GRCm38 87

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream transmembrane emp24 protein transport domain containing 6 Tmed6 1810015P03Rik,
1810018I24Rik
-14.1kb Sense Chr. 8 - 107061484 - 107065644 on NCBI Build GRCm38
Downstream microRNA 7075 Mir7075 Gm27821,
mmu-mir-7075
14.8kb Sense Chr. 8 - 107096025 - 107096106 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800002871.mm.1

Set Membership:

Amplicon length between 86 and 100 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

telomere maintenance
in utero embryonic development
cell cycle
positive regulation of gene expression
negative regulation of gene expression
telomere capping
telomeric loop formation
protection from non-homologous end joining at telomere
regulation of telomere maintenance
negative regulation of telomere maintenance
positive regulation of telomere maintenance
positive regulation of nitric-oxide synthase activity
protein localization to chromosome, telomeric region
negative regulation of beta-galactosidase activity
negative regulation of cellular senescence
DNA binding
chromatin binding
double-stranded telomeric DNA binding
protein binding
protein C-terminus binding
telomeric DNA binding
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800022146.mm.1 NR_104410.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800022146.mm.1 TC0800002871.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0