This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04411028_g1
See other EIF4E1B GE Assays ›
Gene Symbol:
Eif4e1b
Gene Name:
eukaryotic translation initiation factor 4E family member 1B
Gene Aliases:
Eif4eloo, Gm273
Chromosome Location:
Chr.13: 54783927 - 54788459 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04411028_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif4e1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_104406.1 5 - 6 520 69
GenBank mRNA DQ279478.1 - 5 - 6 515 69
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DV658037.1 - 5 - 6 515 Chr. 13 - 54784003 - 54787384 [+] on Build GRCm38 69

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream predicted gene 16249 Gm16249 OTTMUSG00000032672 -3.5kb Anti-sense Chr. 13 - 54777514 - 54783091 on NCBI Build GRCm38
Downstream tetraspanin 17 Tspan17 2210021G21Rik,
AI047581,
Fbxo23,
Tm4sf17
2.4kb Sense Chr. 13 - 54789352 - 54796896 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300000637.mm.1

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

translation
translational initiation
RNA binding
translation initiation factor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300004124.mm.1 NM_001033269.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300004124.mm.1 NM_001039683.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300004124.mm.1 NM_001286178.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300004124.mm.1 NM_001286179.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300004124.mm.1 NM_001286180.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300004124.mm.1 NR_104406.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300004124.mm.1 NR_104407.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300004124.mm.1 TC1300000637.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0