This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04410586_g1
See other MEA1 GE Assays ›
Gene Symbol:
Mea1
Gene Name:
male enhanced antigen 1
Gene Aliases:
Mea-1
Chromosome Location:
Chr.17: 46679138 - 46683126 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04410586_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mea1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_102392.1 1 - 2 181 85
GenBank mRNA AK010863.1 - 1 - 2 59 85
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CN840293.1 - 179 Chr. - - [-] on Build 85
GenBankmRNA CA463489.1 - 1 - 2 98 Chr. 17 - 46681072 - 46682975 [+] on Build GRCm38 85
GenBankmRNA CN834313.1 - 1 - 2 124 Chr. 17 - 46681043 - 46682804 [+] on Build GRCm38 85
GenBankmRNA AI527686.1 - 1 - 2 86 Chr. 17 - 46681081 - 46681837 [+] on Build GRCm38 85
GenBankmRNA CJ145830.1 - 1 - 2 94 Chr. 17 - 46681073 - 46682083 [+] on Build GRCm38 85
GenBankmRNA W29549.1 - 75 Chr. - - [-] on Build 85
GenBankmRNA CJ053938.1 - 1 - 2 79 Chr. 17 - 46681089 - 46682088 [+] on Build GRCm38 85
GenBankmRNA CJ150700.1 - 1 - 2 91 Chr. 17 - 46681078 - 46682820 [+] on Build GRCm38 85
GenBankmRNA BY079410.1 - 1 - 2 59 Chr. 17 - 46681109 - 46682044 [+] on Build GRCm38 85
GenBankmRNA AA118399.1 - 1 - 2 62 Chr. 17 - 46681105 - 46682837 [+] on Build GRCm38 85
GenBankmRNA CJ143258.1 - 1 - 2 60 Chr. 17 - 46681110 - 46682105 [+] on Build GRCm38 85

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream kelch domain containing 3 Klhdc3 1300011D16Rik,
Peas
-56.0 Anti-sense Chr. 17 - 46674550 - 46680930 on NCBI Build GRCm38
Downstream protein phosphatase 2,
regulatory subunit B',
delta
Ppp2r5d B'delta,
TEG-271,
Tex271
1.1kb Anti-sense Chr. 17 - 46682991 - 46705020 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK010863.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s)

multicellular organism development
spermatogenesis
cell differentiation

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700008616.mm.1 NM_010787.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700008616.mm.1 NR_102392.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700008616.mm.1 TC1700000900.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0