This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04410548_m1
See other FANCL GE Assays ›
Gene Symbol:
Fancl
Gene Name:
Fanconi anemia, complementation group L
Gene Aliases:
2010322C19Rik, AW554273, B230118H11Rik, Phf9, Pog, gcd
Chromosome Location:
Chr.11: 26387084 - 26471883 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04410548_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fancl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_102382.1 9 - 10 804 88
GenBank mRNA CN455644.1 - 8 - 9 590 88

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream RIKEN cDNA 5730522E02 gene 5730522E02Rik -258.1kb Anti-sense Chr. 11 - 25616846 - 26210576 on NCBI Build GRCm38
Downstream vaccinia related kinase 2 Vrk2 2810003O05Rik,
AI447698
1.6kb Anti-sense Chr. 11 - 26471401 - 26594014 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100000264.mm.1

Set Membership:

Amplicon length between 86 and 100 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of defense response to virus by host
DNA repair
protein monoubiquitination
cellular response to DNA damage stimulus
gamete generation
protein ubiquitination
regulation of cell proliferation
ubiquitin-protein transferase activity
protein binding
zinc ion binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
ubiquitin protein ligase binding
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
metal ion binding
ubiquitin protein ligase activity
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100002270.mm.1 NM_001277273.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100002270.mm.1 NM_025923.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100002270.mm.1 NR_102382.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100002270.mm.1 TC1100000264.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100002272.mm.1 NM_001277273.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100002272.mm.1 NM_025923.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100002272.mm.1 NR_102382.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100002272.mm.1 TC1100000264.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100002269.mm.1 NM_001277273.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100002269.mm.1 NM_025923.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100002269.mm.1 NR_102382.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100002269.mm.1 TC1100000264.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100002262.mm.1 NM_001277273.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100002262.mm.1 NM_025923.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100002262.mm.1 NR_102382.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100002262.mm.1 TC1100000264.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100002267.mm.1 NM_001277273.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100002267.mm.1 NM_025923.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100002267.mm.1 NR_102382.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100002267.mm.1 TC1100000264.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0