This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04394036_g1
See other ACTB GE Assays ›
Gene Symbol:
Actb
Gene Name:
actin, beta
Gene Aliases:
Actx, E430023M04Rik, beta-actin
Chromosome Location:
Chr.5: 142903116 - 142906724 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04394036_g1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Actb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK075973.1 - 1 - 2 69 72
AK078935.1 - 1 - 2 72 72
AK088691.1 - 1 - 2 69 72
AK145191.1 - 1 - 2 72 72
AK145196.1 - 1 - 2 72 72
AK145308.1 - 1 - 2 72 72
AK147787.1 - 1 - 2 69 72
AK149920.1 - 1 - 2 72 72
AK150545.1 - 1 - 2 72 72
AK150553.1 - 1 - 2 72 72
AK150558.1 - 1 - 2 72 72
AK150570.1 - 1 - 2 72 72
AK150662.1 - 1 - 2 69 72
AK150711.1 - 1 - 2 72 72
AK150854.1 - 1 - 2 72 72
AK150866.1 - 1 - 2 72 72
AK150879.1 - 1 - 2 72 72
AK151010.1 - 1 - 2 69 72
AK151136.1 - 1 - 2 72 72
AK151145.1 - 1 - 2 69 72
AK151159.1 - 1 - 2 72 72
AK151166.1 - 1 - 2 69 72
AK151190.1 - 1 - 2 72 72
AK151202.1 - 1 - 2 72 72
AK151226.1 - 1 - 2 69 72
AK151277.1 - 1 - 2 69 72
AK151297.1 - 1 - 2 72 72
AK151350.1 - 1 - 2 69 72
AK151385.1 - 1 - 2 72 72
AK151386.1 - 1 - 2 69 72
AK151398.1 - 1 - 2 72 72
AK151444.1 - 1 - 2 72 72
AK151468.1 - 1 - 2 69 72
AK151663.1 - 1 - 2 72 72
AK151712.1 - 1 - 2 73 72
AK151898.1 - 1 - 2 69 72
AK151956.1 - 1 - 2 68 72
AK151995.1 - 1 - 2 72 72
AK151999.1 - 1 - 2 72 72
AK152043.1 - 1 - 2 72 72
AK152052.1 - 1 - 2 69 72
AK152083.1 - 1 - 2 72 72
AK152218.1 - 1 - 2 72 72
AK152433.1 - 1 - 2 69 72
AK152470.1 - 1 - 2 69 72
AK152591.1 - 1 - 2 72 72
AK152615.1 - 1 - 2 72 72
AK152651.1 - 1 - 2 72 72
AK152671.1 - 1 - 2 72 72
AK152813.1 - 1 - 2 72 72
AK152841.1 - 1 - 2 72 72
AK152844.1 - 1 - 2 69 72
AK153461.1 - 1 - 2 69 72
AK159759.1 - 1 - 2 72 72
AK159834.1 - 1 - 2 69 72
AK160029.1 - 1 - 2 69 72
AK166349.1 - 1 - 2 72 72
AK166498.1 - 1 - 2 72 72
AK167117.1 - 1 - 2 72 72
AK167825.1 - 1 - 2 72 72
AK167960.1 - 1 - 2 72 72
BY783897.1 - 1 - 2 100 72
J04181.1 - 1 - 2 72
X03672.1 - 1 - 2 71 72

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK088691.1, AK145191.1, AK145196.1, AK145308.1, AK147787.1, AK150545.1, AK150553.1, AK150558.1, AK150711.1, AK150854.1, AK150866.1, AK150879.1, AK151010.1, AK151136.1, AK151145.1, AK151159.1, AK151166.1, AK151190.1, AK151202.1, AK151226.1, AK151277.1, AK151297.1, AK151350.1, AK151385.1, AK151386.1, AK151398.1, AK151444.1, AK151468.1, AK151663.1, AK151898.1, AK151956.1, AK151995.1, AK151999.1, AK152043.1, AK152052.1, AK152083.1, AK152218.1, AK152433.1, AK152470.1, AK152591.1, AK152615.1, AK152651.1, AK152844.1, AK153461.1, AK159759.1, AK159834.1, AK160029.1, AK166349.1, AK166498.1, AK167117.1, AK167825.1, AK167960.1, J04181.1, X03672.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

Panther Classification:

Molecular Function -

actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 608492 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0