This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04336209_s1
See other SNORA7A GE Assays ›
Gene Symbol:
Snora7a
Gene Name:
small nucleolar RNA, H/ACA box 7A
Gene Aliases:
MBI-141
Chromosome Location:
Chr.6: 115807975 - 115808103 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04336209_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Snora7a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_028546.1 1 82 125
GenBank mRNA AF357398.1 - 1 82 125
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DT918153.1 - 1 327 Chr. 6 - 115807111 - 115808348 [-] on Build GRCm38 125

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ribosomal protein L32 Rpl32 AU020185,
rpL32-3A
Sense Chr. 6 - 115805514 - 115808743 on NCBI Build GRCm38
Downstream EF-hand calcium binding domain 12 Efcab12 BB124106 2.6kb Sense Chr. 6 - 115810729 - 115838791 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10547074 - -
Affymetrix Mouse Gene 1.1 ST Array 10547073 - -
Affymetrix Mouse Clariom S Array PSR0600022750.mm.1 NR_028546.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600022750.mm.1 NR_028546.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600022750.mm.1 TC0600002961.mm.1 MTA TC
Murine Genome U74v2 Set 99198_PM_at - -
Murine Genome U74v2 Set 99198_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0