This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm04335780_m1
See other ELP3 GE Assays ›
Gene Symbol:
Elp3
Gene Name:
elongator acetyltransferase complex subunit 3
Gene Aliases:
2610507P14Rik, KAT9
Chromosome Location:
Chr.14: 65530446 - 65593112 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm04335780_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Elp3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_045599.1 14 - 15 1827 85
GenBank mRNA AK150058.1 - 14 - 15 1765 85

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream RIKEN cDNA 9530047P18 gene 9530047P18Rik -63.4kb Anti-sense Chr. 14 - 65465797 - 65467093 on NCBI Build GRCm38
Downstream nuclear GTPase,
germinal center associated
Nuggc Gm600,
SLIP-GC
46.3kb Anti-sense Chr. 14 - 65593499 - 65648444 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400002259.mm.1

Set Membership:

Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

neuron migration
tRNA wobble uridine modification
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
nervous system development
central nervous system development
histone acetylation
positive regulation of cell migration
histone H3 acetylation
histone H4 acetylation
regulation of protein kinase activity
RNA polymerase II core binding
catalytic activity
histone acetyltransferase activity
N-acetyltransferase activity
phosphorylase kinase regulator activity
H3 histone acetyltransferase activity
H4 histone acetyltransferase activity
transferase activity
transferase activity, transferring acyl groups
metal ion binding
iron-sulfur cluster binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400017398.mm.1 NM_001253812.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017398.mm.1 NM_001253812.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017398.mm.1 NM_028811.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017398.mm.1 NM_028811.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400017398.mm.1 NR_045599.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017398.mm.1 NR_045599.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400017398.mm.1 TC1400002259.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0